Corning® Cell Counter, Corning
Supplier: Corning
6749
76200-994EA 4589.31 USD
76200-994
Corning® Cell Counter, Corning
Cell Counters and Flow Cytometers Electronic Cell Counters
This cell counter uses online image processing with a cloud-based machine learning algorithm.
  • Fast and accurate results: Fits into the palm of a hand
  • Cell count in less than 3 seconds
  • Sophisticated deep neural network for cell detection with optimal accuracy – when Trypan blue is added, the system can also detect cell viability
Connect to a Windows® 10 computer or tablet and start the Cytosmart Cloud app. Place the counting chamber on stage, focus on the cells, and press the counting function.
The simplicity of the device allows anyone working inMore Product Information