Workbrutes® PVC Overshoes, Tingley Rubber
Supplier: Tingley Rubber
35111-2X 35111-XS
76199-154EA 12.34 USD
76199-154 76199-156 76199-158 76199-744 76199-746 76199-748
Workbrutes® PVC Overshoes, Tingley Rubber
Shoes and Boots
PVC Overshoes with thicker, deeper cleats for better grip and longer wear.
  • Covers work shoe up to ankle
  • Heavy duty heel kick off for easy off
Frigiflex® PVC compound contains special additives for high stretch and flexibility in cold temperatures. The seamless construction provides 100% waterproof protection.
Cleated outsole spits out debris and provides excellent traction. The injection molding puts less material in the upper, eliminating unnecessary weight, for reduced leg fatigue. Engineered for a longer life by adding more material to the soleMore Product Information