Cap Strips for Well Plates, Amplitube™, Simport
Supplier: Simport Scientific
T323-1N
76198-306PK 26.95 USD
76198-306
Cap Strips for Well Plates, Amplitube™, Simport
Seals Microplate Seals
Amplitube™ domed cap strips ensure a perfect closure and a positive seal during thermal cycling and prevent evaporation.
  • 8 domed cap strips
  • Made of FDA compliant polypropylene
  • Free of RNase, DNase, Pyrogen, and DNA
  • Leakproof/evaporation tested
Packaging: Packaged in tamperproof resealable bags.