LaboPlast® and SteriPlast® DispoDipper, Burkle
Supplier: BURKLE INC
5393-7001 5393-7000 5393-7010 5393-7011 5393-7020 5393-7021
76183-094PK 67.94 USD
76183-094 76183-154 76299-420 76299-432 76299-434 76299-436
LaboPlast® and SteriPlast® DispoDipper, Burkle
Samplers Disposable Samplers
This sampling tube is manufactured in a cleanroom specially designed for disposable use. Due to the blue colour, it is also perfect for use in the food, animal feed, and pharmaceutical industries. The scaled sample container can be used for an exact measurement. After sampling, the container can be closed and is leakproof.
  • Polypropylene, blue transparent
  • Individually packaged and optionally sterilized by gamma rays
  • Production and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
  • Scaled sampling tube
  • Immersion
More Product Information