PIPETMAN® Expert Tipack™ Racked Tips, Gilson®
Supplier: GILSON, INC.
F1731002 F1733012 F1736002 F1736012 F1732002 F1731011 F1733011 F1736011 F1732012 F1733001 F1736001 F1732001 F1735001 F1735011 F1735002 F1735012 F1731012 F1733002 F1732011 F1731001
76178-312CS 366.59 USD
76178-312 76178-318 76178-320 76178-326 76178-338 76178-342 76178-344 76178-346 76178-386 76178-438 76178-440 76178-442 76178-456 76178-468 76180-036 76180-040 76180-042 76180-368 76180-440 76180-446
PIPETMAN® Expert Tipack™ Racked Tips, Gilson®
Pipette Tips
Universal fit PIPETMAN® EXPERT Tips are designed to fit leading pipette brands, allowing you to stock one brand of tips saving time, money, and bench space. The tips are produced using state-of-the-art manufacturing techniques assuring lot-to-lot consistency and high performance. Volume graduations make it easy to visually check the volume aspirated. Color-coded packaging corresponds to industry standards for color matching to respective Gilson pipette models.
  • Universal fit
  • Free of detectable contaminants
  • Convenient, economical packaging
Universal fitMore Product Information