PIPETMAN® L Fixed Volume Single Channel Pipettor, Gilson®
Supplier: GILSON, INC.
FA10030 FA10024 FA10020 FA10017 FA10025 FA10021 FA10026 FA10022 FA10018 FA10027 FA10028 FA10031 FA10029 FA10023 FA10019
76178-098EA 430.22 USD
76178-098 76178-406 76178-408 76178-410 76178-412 76178-414 76178-416 76178-418 76178-420 76178-422 76178-424 76178-426 76178-428 76178-430 76178-432
PIPETMAN® L Fixed Volume Single Channel Pipettor, Gilson®
Pipettes Micropipettes
The PIPETMAN® L Fixed pipettes offer the same comfortable, lightweight design as the variable volume models with a fixed volume that reduces the risk of any volume error during pipetting. This makes them an ideal choice for clinical diagnostics, quality control, and any routine testing applications even for non-trained lab technicians.
  • Comfortable pipetting
  • GLP compliance
Based on the ergonomic design of the original PIPETMAN L, the fixed volume model offers an ideal weight and handle shape. The more natural fit of the pipette lets you relax your grip,More Product Information