PIPETMAN® L Single Channel Pipettor, Gilson®
Supplier: GILSON, INC.
FA10005M FA10006M FA10007 FA10008 FA10001M FA10002M FA10003M FA10004M
76178-052EA 662.28 USD
76178-052 76178-054 76178-056 76178-058 76178-348 76178-350 76178-352 76178-354
PIPETMAN® L Single Channel Pipettor, Gilson®
Pipettes Micropipettes
Designed for performance and comfort, PIPETMAN® L comes in a comprehensive range of models, adapted to your needs in the lab. PIPETMAN L air-displacement pipettes cover a full volume range. Comfortable, precise, accurate, and reliable, use PIPETMAN L pipettes confidently in your applications.
  • Convenient use
  • Secure pipetting
  • GLP compliance
Lightweight and balanced, PIPETMAN L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue. Right-handed or left-handed? No problem. Adjust theMore Product Information