Trichloroacetic acid (TCA) solution 6.1 N

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

0219605780 0219605790
76177-482EA 92.46 USD
76177-482 76177-484
Trichloroacetic acid (TCA) solution 6.1 N
Trichloroacetic acid (TCA)

Trichloroacetic acid is an acetic acid analogue commonly used to precipitate proteins, DNA and RNA. In the presence of SDS Trichloroacetic acid will precipitate proteins which can then be quantified by the Lowry method. The compound has also found uses as a decalcifier and fixative in microscopy, in protein sequencing, detecting albumin and organic synthesis.
Trichloroacetic acid is used in protein precipitation; has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning. Rats exposed to chronic sublethal amounts of trichloroacetic acid displayed an increase in serum bilirubin with a decrease in hematological proteins and cholesterols along with significant decreases in red blood corpuscles, mean cell volume, mean corpuscular hemoglobin, hemoglobin and hematocrit. In mice the production of liver tumors was associated to Trichloroacetic acid, which is also a known mouse hepatocarinogen. On the guinea pig trichloroacetic acid was observed to be a mild alergen on the skin.
+4 °C


  • Concentration: 6.1N Solution
  • Presentation: Solution
  • Vapor Density: 5.6 (air = 1)(Lit.)
  • Vapor Pressure: 1.2 hPa at 50 °C(Lit.)
  • pH: 1.2 (0.1 M aqueous solution)(Lit.)

Formula: Cl₃CCOOH
Boiling Pt: 197 °C (1013 hPa) (lit.)
Melting Pt: ∼56 °C (lit.)
Density: 1.61 g/cm³ (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2564
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR