Ribavirin

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide

0219606650 0219606610
76177-334EA 199.28 USD
76177-334 76177-488
Ribavirin
Ribavirin

Ribavarin is an antiviral compound reported to be active against RSV, HIV, HCV and other infectious agents. Its metabolite, ribavirin 5'-phosphate, inhibits inosine monophosphate dehydrogenase (IMP). It is also an analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.
Ribavirin is active against a number of DNA and RNA viruses. It is a member of the nucleoside antimetabolite drugs that interfere with duplication of viral genetic material.
Antiviral agent used against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. Its metabolite, ribavirin 5′-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase, but many other mechanisms of action are also supported with experimental evidence.
Antiviral agent whose metabolite, ribavirin 5′-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase. Used to inhibit purine (GTP pools) biosynthesis at the level of inosine monophosphate (IMP) dehydrogenase and as a precursor to the di- and tri-phosphates which inhibit viral RNA-dependent RNA polymerases.
Store at +4 °C.


  • Presentation: White Powder
  • Soluble in water (20 mg/mL - clear, colorless solution).

Formula: C8H12N4O5
Melting Pt: 173 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00058564
CAS Number: 36791-04-5

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
Specific Rotation Between -33.5° and -37.0°
pH 4.0-6.5
Loss on Drying ≤0.5%
Residue on Ignition ≤0.25%
Heavy Metals ≤0.001%
Chromatographic Purity Individual Peak ≤0.25%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR