Anti-MMP-14 Polyclonal Antibody

Supplier: Boster Biological Technology
PB9283
76173-400EA 702.73 USD
76173-400
Anti-MMP-14 Polyclonal Antibody
Antibodies
Polyclonal antibody for MT1 MMP/MMP14 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. MT1 MMP/MMP14 information: Molecular Weight: 65894 MW; Subcellular Localization: Membrane ; Single-pass type I membrane protein . Melanosome. Cytoplasm. Identified by mass spectrometry in melanosome fractions from stage I to stage IV. Forms a complex with BST2 and localizes to the cytoplasm; Tissue Specificity: Expressed in stromal cells of colon, breast, and head and neck. Expressed in lung tumors.

Type: Primary
Antigen: MMP-14
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR