Anti-TFRC Polyclonal Antibody

Supplier: Boster Biological Technology
PB9233
76173-332EA 787.77 USD
76173-332
Anti-TFRC Polyclonal Antibody
Antibodies
Polyclonal antibody for TRANSFERRIN RECEPTOR/TFRC detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. TRANSFERRIN RECEPTOR/TFRC information: Molecular Weight: 84871 MW; Subcellular Localization: Cell membrane ; Single-pass type II membrane protein . Melanosome . Identified by mass spectrometry in melanosome fractions from stage I to stage IV.

Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR