Serum Free Cryo Medium, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
163-320-051
76169-066EA 88.52 USD
76169-066
Serum Free Cryo Medium, Quality Biological
Cell Culture Media
A sterile, 2X concentrated freezing medium containing DMSO developed for the cryopreservation of cells cultured in serum free conditions.
  • Animal-free, protein-free, chemically defined cryopreservation medium Without phenol red
  • Contains DMSO and other cryoprotective components
  • 2X concentration
  • Absence of FBS allows continuous culture and preservation of cells under serum free condition
Serum Free Cryo Medium's 2X concentrated formulation allows for dilution with either conditioned medium or complete fresh medium. Serum Free Cryo Medium can be easily paired with serum free medium, QBSF-55 ( #10128-230) for culturing cells in serum free conditions.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: Accesory not included.
Caution: Research Use Only