HYPO-CHLOR® Sodium Hypochlorite Neutral, Veltek
Supplier: Veltek Associates
SHC-NPH-0.25-02 SHC-NPH-0.52-16Z SHC-NPH-0.52-02 SHC-NPH-0.25-16Z
76102-184CS 361.7 USD
76102-184 76102-186 76102-188 76102-190
HYPO-CHLOR® Sodium Hypochlorite Neutral, Veltek
Cleaning Agents Disinfectants
HYPO-CHLOR® Neutral Products are a ready-to-use and neutralized 0.52% and 0.25% sodium hypochlorite solutions formulated with Water for Injection (WFI).
  • Filled in an ISO 5 (Grade A/B, Former Class 100)
  • Filtered at 0.2 microns
  • Components are air washed with 0.2 micron filtered air
  • Aseptically filled into sterile components via gamma irradiation
  • Lot sterility tested according to current USP compendium
  • Completely traceable from start to finish
  • Safe to use daily on stainless steel surfaces
Designed for all pharmaceutical, biopharmaceutical, biotechnology,More Product Information