EZwaste™ XL Replacement Chemical Exhaust Filter, Foxx Life Sciences
Supplier: VWR International
76018-680 76018-682
76018-680PK 839.86 USD
76018-680 76018-682
EZwaste™ XL Replacement Chemical Exhaust Filter, Foxx Life Sciences
Filters Liquid Process Filters
The Ezwaste™ XL Solvent Waste Filter is designed to be an affordable solution for HPLC liquid waste disposal. The activated carbon filter traps harmful VOCs from being released into the air.
  • Affordable solution for HPLC waste disposal
  • Activated carbon filter traps harmful VOCs
  • Closed loop design
  • Breakthrough indicators available
The EZwaste™ XL Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal.
Depending on application, carbon exhaust filter life cycle may vary. Filters are supplied in packs of 2.