PAL SPME Arrows and Kits, Restek
Supplier: Restek
27490 27491 27492 27493 27494 27498 27499 27500 27501 27495 27496 27497 27502 27503 27504 27505 27506 27507 27508
75996-520EA 660.79 USD
75996-520 75996-522 75996-524 75996-526 75996-528 75996-530 75996-532 75996-534 75996-536 75996-538 75996-540 75996-542 75996-544 75996-546 75996-548 75996-550 75996-552 75996-554 75996-556 75996-558 75996-560 75996-562 75996-564 75996-566 75996-568
PAL SPME Arrows and Kits, Restek
Chromatography Supplies Chromatography Spare Parts
The PAL SPME Manual Injection Kit is designed to house SPME Arrows and traditional SPME fibers during extraction and injection steps.
  • Rugged stainless steel construction
  • Longer lifetime
  • Faster extraction
  • Higher sample throughput
  • Better sensitivity allows lower LODs
Due to the relatively large diameter of Restek PAL SPME Arrows, you must modify the GC inlet using an instrument-specific conversion kit prior to use.
Accessories information: A GC inlet conversion kit is required in order to use a Restek PAL SPME Arrow.
Ordering information: TheseMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable