Aluminium Weighing Dishes, Qorpak
Supplier: Qorpak
275591
75993-628CS 569.06 USD
75993-628
Aluminium Weighing Dishes, Qorpak
Boats Weighing Boats
General-purpose disposable aluminum weighing dishes can be used as evaporating dishes, dust covers, or balance pans.
  • Ideal for dispensing, sampling, and mixing
These crimped wall weigh dishes have no oil residue or vinyl coating. They offer feature increased rigidity and a large tab. Great for routine weighing and moisture testing, as well as an embedding mold.