solarus® Digital Bottle-Top Burets, Hirschmann
Supplier: Hirschmann
9392020 9392050 9392010
83007-660EA 2013.63 USD
83007-660 83007-662 75993-418
solarus® Digital Bottle-Top Burets, Hirschmann
Burets
Ideal for manual titrations in analytical labs, water and wastewater plants, and QA processes.
Integrated solar panels power the electronic components and the display, eliminating the need for charging or changing batteries
Exchangeable valves and simple user-specific Quick-Cal calibration make individual adaptations possible. However, the system also has a permanent default setting at 20°C (68°F) with water. A serial reagent recirculation system ensures quick, simple priming and prevents the waste of reagents. A button on the display will switch from the default calibration to theMore Product Information