Anti-C3 Goat Polyclonal Antibody (Fluorescein)

Supplier: MP Biomedicals
0855385
75983-048EA 482.17 USD
75983-048
Anti-C3 Goat Polyclonal Antibody (Fluorescein)
Antibodies
The complement system provides innate defense against microbial infection and is a "complement" to antibody mediated immunity. The complement system consists of thirty-five interacting plasma and membrane associated proteins which contribute to host-defense and initiate and amplify inflammation, even in the preimmune state where specific antibodies and lymphocytes are not available. In addition to the complement components themselves, this system also contains several soluble factors that prevent spontaneous complement activation from occurring in solution, as well as, several regulatoryMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol C3
Antigen Name Complement C3
Environmentally Preferable
Antibody Type Primary
Clonality Polyclonal
Conjugation Fluorescein
Reactivity Guinea pig
Host Goat
Isotype
ImmunoChemistry Yes
ImmunoFluorescence Yes
Form Lyophilized
Storage Buffer 7.3
Shipping Temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR