Plastic Transfer Pipettes, Argos Technologies
Supplier: Antyila Scientific
06226-11 06226-12 06226-13 06226-15 06226-17 06226-19 06226-21 06226-22 06226-23 06226-24 06226-25 06226-26 06226-01 06226-03 06226-27 06226-28 06226-29 06226-30 06226-31 06226-32
75982-354BX 12.76 USD
75982-354 75982-351 75982-353 75982-355 75982-357 75982-359 75982-361 75982-363 75982-365 75982-367 75982-369 75982-371 75982-373 75982-407 75982-409 75982-411 75982-413 75982-415 75982-417 75982-419 75982-421 75982-423
Plastic Transfer Pipettes, Argos Technologies
Pipettes Pasteur/Transfer Pipettes
Perfect for single use fluid transfers.
  • Can be frozen in liquid nitrogen
  • Temperature range of –50 to 90 °C
  • Can be gas sterilized
  • Plastic reduces cell loss and protein loss, which often occurs with binding that takes place with glass alternatives
  • Pipettes can be heat sealed to provide liquid tight transport, or storage of solutions
Argos Technologies transfer pipettes are manufactured from a custom grade low-density polyethylene (LDPE), making them the perfect substitute for breakable glass pipettes.
These single unit pipettes are disposable,More Product Information