Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: HPCA1/763]
Supplier: Biotium
BNCB0763-100 BNCB0763-500
75894-334EA 205.77 USD
75894-334 75894-336
Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: HPCA1/763]
Antibodies
This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120 kDa, which is identified as CD34. Its expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, CD34 is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. Its expression is also found in vascular endothelium. Additionally, it appears that proliferatingMore Product Information