Pro20i Dissolved Oxygen Instrument, YSI
Supplier: YSI
607133 607130 607131 607125 607129 607132
75881-062EA 1346.97 USD
75881-062 75881-078 75881-080 75881-082 75881-084 75881-086
Pro20i Dissolved Oxygen Instrument, YSI
Electrochemistry Meters Oxygen Meters
Dissolved oxygen and temperature handheld instrument with integral (i.e. non-detachable) cable. The Pro20i is a direct replacement for the model 55 and 550A dissolved oxygen instruments.
  • Integral (i.e. built-in) cable with 1, 4, 10 m field cable; user-replaceable DO sensors
  • One Touch Cal allows for easy DO calibrations within 3 seconds with the press of a button.
  • Automatic barometric pressure compensation
  • Quick response times; 95% response time in approximately 8 seconds with standard membrane
  • Stores 50 data sets; no need to write down data
  • Large graphic,
More Product Information