Topaz GC Inlet Liners for Agilent Gas Chromatographs, Packed, Restek
Supplier: Restek
23432 23303 23314 23316 23305 23284 23300 23307 23309 23467 23468
75877-860PK 613.52 USD
75877-860 75877-876 75877-882 75877-886 75878-238 75878-282 75878-284 75878-288 75878-290 75878-308 75878-310
Topaz GC Inlet Liners for Agilent Gas Chromatographs, Packed, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Order Now
SPECIFICATIONS
Environmentally Preferable