S-100 IHC Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
3740A 3740B
75876-922BX 91.27 USD
75876-922 75876-924
S-100 IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
S-100 control slides contain sections of positive staining melanoma and negative staining lung.
  • Tissue type: (+) Melanoma / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide
Newcomer supply S-100 control slides are for the positive immunohistochemical staining of S-100, a ubiquitous protein expressed in a number of cells