Eppendorf® New Brunswick™ Innova® 44/44R Stackable Incubator Shakers
Supplier: Eppendorf
M1282-0000 M1282-0010 M1282-0004 M1282-0014
75874-524EA 24946.88 USD
75874-524 75874-528 75874-530 75875-646 75875-648
Eppendorf® New Brunswick™ Innova® 44/44R Stackable Incubator Shakers
Shakers and Mixers Shaking Incubators
The Innova 44 and 44R (refrigerated) are Eppendorf’s most popular shakers - they accommodate flasks up to 5 L, have the space-saving glide-up door, and the reliable Innova triple-eccentric drive. They just run forever to give you peace of mind.
  • Applicational flexibility: Accommodates flasks up to 5 L, sticky pad platform for flat bottom vessels, orbits 25 or 51 mm (1 or 2"), optional: photosynthetic light bank and humidity monitoring
  • Saves money and lab space: No internal HEPA-filter that must be replaced annually for optimal performance, 2×/3× stackable
More Product Information