(±)-3-Octanol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol

O1462-500ML
75872-774EA 98.02 USD
75872-774 75872-776 75873-120
(±)-3-Octanol
(±)-3-Octanol

3-Octanol is a fragrance ingredient with an earthy or musty odor.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₈O
Density: 0.818
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR