SP Scientific miVac Sample Holder Jet Rotors for Quattro Concentrators, Genevac
Supplier: Genevac
QRC-01075-236 QRC-01275-200 QRC-13100-100 QRC-15&20EPP-200 QRC-15CCT-072 QRC-15EPP-200 QRC-15V60-116 QRC-16100-100 QRC-17V60-116 QRC-18150-040 QRC-20EPP-200 QRC-25150-032 QRC-25V100-038 QRC-28V140-030 QRC-28V60-042 QRC-28V95-038 QRC-29120-036 QRC-35105-032 QRC-44100-014 QRC-50CCT-036
75871-548EA 2155.55 USD
75871-548 75871-550 75871-552 75871-554 75871-556 75871-558 75871-560 75871-562 75871-564 75871-566 75871-576 75871-578 75871-580 75871-582 75871-584 75871-586 75871-588 75871-590 75871-592 75871-594
SP Scientific miVac Sample Holder Jet Rotors for Quattro Concentrators, Genevac
Centrifuges Rotors
Lightweight, easily interchangeable sample holders for Quattro Concentrators hold a variety of glass and plastic tubes and vials.
  • Lightweight solid aluminum construction
  • Convenient handles
  • Hard-wearing and highly durable
  • Wide selection available
Each miVac rotor is made to exacting specifications to assure long life, secure holding of samples and maximum heat transfer and reliability
Lightweight solid aluminum material has a very high thermal conductivity, allowing for excellent heat transfer. Precise specifications assure tubes fit tightlyMore Product Information