Swann Morton Scalpels, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
SM0501 SM0503 SM0505 SM0506 SM0507 SM0508 SM0510
75870-492BX 41.21 USD
75870-492 75870-494 75870-496 75870-498 75870-500 75870-502 75870-626
Swann Morton Scalpels, Cincinnati Surgical
Scalpels and Scalpel Assemblies Scalpels
Swann Morton is a world leader in the manufacturing of surgical blades, scalpels, and handles
  • 200% visual inspection
  • Packaged with VCI paper to protect the blade from corrosion
  • Strongest and smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Blue handle
It is a name respected globally for quality, precision, consistency, and reliability.
Certifications: CE marked.