VWR® Advanced Dry Block Heaters, 230 V (Export Only)
Supplier: VWR International
80863251 80863259 80863260 80863266 80863267 80863272 80863273 80863256 80863257 80863250
75838-266EA 2099.09 USD
75838-266 75838-274 75838-276 75838-288 75838-290 75838-300 75838-302 75838-320 75838-322 75838-324
VWR® Advanced Dry Block Heaters, 230 V (Export Only)
Block Heaters Digital Block Heaters
Designed for applications that require repeatable results and superior temperature stability.
Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures
230 V model, not for use in USA
New design with smaller footprint
Exceptional temperature uniformity and stability
Heated lid model reduces condensation in sample lids
LED display for temperature and time
Heats to 120 °C (245 °F)
Holds interchangeable, modular heating blocks
5 year warranty on parts and labor
Available with a NISTMore Product Information