BioFit Urethane Lab Chairs and Stools
Supplier: VWR International
75821-332 UUA-H-RC-C-ATF 75821-336 75821-338 75821-340 UUA-M-HG-C-ATF UUA-M-RC-C-ATF 75821-346 UUS-H-RC-C 75821-350 75821-352 75821-354
75821-332EA 1309.44 USD
75821-332 75821-334 75821-336 75821-338 75821-340 75821-342 75821-344 75821-346 75821-348 75821-350 75821-352 75821-354
BioFit Urethane Lab Chairs and Stools
Chairs
Lab Chairs and stools are made of urethane, crafted with user comfort, health and wellbeing in mind.
  • Easy-to-clean seats and backrests that resist water, chemicals, paints and more
  • Easily adjust seat-height from seated position
  • Height and tilt adjustable backrests with integrated lumbar support
  • Dual-wheel resistance casters enhance user safety by inhibiting seating from rolling away on hard floors or 2" high polyamide glides
  • Adjustable and fixed height footrings
BioFit seating solutions are crafted with user comfort, health and wellbeing in mind