MLA D-Tipper Pipettors, VistaLab
Supplier: VistaLab
1141C 1150C 1163C 1159C 1148C 1157C 1051C 1054C 1058C 1059C 1060C 1143C 1154C 1162C 1053C 1056CLT 1142C 1146C 1152C 1144C 1149C 1055C 1056C 1057C 1132 1153C 1051PK 1052PK 1054PK 1055PK 1056PK 1057PK 1059PK 1060PK 1147C 1151C 1160C 1050-0998 1050-0999 1052C
49000-536EA 447.12 USD
49000-536 53514-092 53514-100 49000-538 49000-542 49000-544 53514-026 53514-032 53514-042 53514-044 53514-046 53514-088 53514-098 49000-540 53514-030 53514-038 53514-086 53514-090 53514-094 49000-534 49000-546 53514-034 53514-036 53514-040 53514-076 53514-096 89368-660 89368-662 89368-664 89368-666 89368-668 89368-670 89368-672 89368-674 75813-772 75814-370 75814-374 75814-388 75814-390 75814-392
MLA D-Tipper Pipettors, VistaLab
Pipettes Repetitive Pipettors
MLA pipettors feature a built-in tip ejector and single-stroke 'to deliver' operation.
  • Glove guard
  • Available in a wide range of fixed volumes
  • Light plunger forces for easy operation with thumb/forefinger
  • Built-in tip ejector
  • Individually serialized
  • Lifetime warranty on parts
MLA pipettes are made of precision-engineered aluminum alloy and stainless steel.
These pipettors are available in 30 fixed volumes ranging from 5 to 1000 μl, or as a selectable model that is compatible at volumes of 50, 100 and 200 μl. Pipettors feature color-coded bodies forMore Product Information