Ethanol absolute ≥99.5%, BiotechGrade (200 Proof)

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethyl alcohol

E1424-4LT E1424-500ML
75811-066EA 787.96 USD
75811-066 75811-068
Ethanol absolute ≥99.5%, BiotechGrade (200 Proof)
Ethanol

Dehydrated Alcohol, 200 Proof, BiotechGrade The GR is an EMD trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.


GHS hazard : Flame;Health hazard;Exclamation mark
UN1170

Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3
Melting Pt: -117
Density: 0.794
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3 II
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (By Volume) 99.5 %
Dnase and Rnase Non-Detected
Water max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR