Surgipath® Snowcoat® Beveled-Edge Microscope Slides, Leica
Supplier: Leica Biosystems
3800373
75809-504CS 452.8 USD
75809-504
Surgipath® Snowcoat® Beveled-Edge Microscope Slides, Leica
Slides Microscope Slides
Snowcoat Beveled Edge Slides feature beveled edges and corners that make for easier handling.
  • Beveled edges
  • Easy handling
  • For blood smears
Slides are good for blood smears, producing a feathered edge. Painted white surface facilitates quality marking adherence. Each slide is washed and carefully inspected to ensure quality.