Biox Tri-Cornered Beakers
Supplier: TWD Scientific
BX93-0050 BX93-0100 BX93-0250 BX93-0400 BX93-1000
75799-910CS 47.08 USD
75799-910 75799-912 75799-914 75799-916 75799-918
Biox Tri-Cornered Beakers
Beakers
Tri-Cornered Beakers are designed to provide three dripless pouring spouts
  • Strong medical-grade polypropylene construction
  • Equipped with three dripless pouring spouts
  • Good impact strength
  • Translucent and autoclavable
  • Disposable, easy to clean, and durable enough for reuse
  • Molded Graduations
Available in six different sizes.
Order Now
SPECIFICATIONS
Environmentally Preferable