Steri-Drape™ Hand Drape, 3M™
Supplier: Healthcare Products
MMM 9052
75787-292CS 1268.54 USD
75787-292
Steri-Drape™ Hand Drape, 3M™
First Aid Supplies Surgical Drapes
All-in-one T-shaped drape covers the entire patient and arm board.
  • One-piece design with integral patient and arm board cover provides quick and easy draping
  • Absorbent fabric prevents fluid strikethrough, making double layering unnecessary
Three tube organizers keeps operative site clear of tubes and cords. Elastomeric aperture isolates the operative site at the hand/arm.