Salmonella O Group Antisera, Pro-Lab Diagnostics
Supplier: PRO-LAB DIAGNOSTIC
PLA021 PLA022 PLA023 PLA024 PLA025 PLA026 PLA027 PLA028 PLA029 PLA030 PLA031 PLA032 PLA033 PLA034 PLA035 PLA036 PLA037 PLA038 PLA039 PLA040 PLA041 PLA042 PLA043 PLA044 PLA045 PLA046 PLA047 PLA048 PLA049 PLA051 PLA052 PLA053 PLA054 PLA055 PLA056 PLA057 PLA058 PLA059 PLA060 PLA061 PLA062
75784-978EA 389.14 USD
75784-978 75784-980 75784-982 75785-224 75785-226 75785-228 75785-230 75785-232 75785-234 75785-236 75785-238 75785-240 75785-242 75785-244 75785-246 75785-248 75785-250 75785-252 75785-254 75785-256 75785-258 75785-260 75785-262 75785-264 75785-266 75785-268 75785-270 75785-272 75785-274 75785-276 75785-278 75785-280 75785-282 75785-284 75785-286 75785-288 75785-290 75785-292 75785-294 75785-296 75785-298
Salmonella O Group Antisera, Pro-Lab Diagnostics
Microbiology Test Systems Antibiotic Tests
Salmonella O antisera is specific for slide agglutination testing.
  • Shelf lives up to five years from date of manufacture
  • Supplied in color coded dropper bottles
The range includes a full panel of polyvalent somatic antisera, monovalent somatic antisera, polyvalent flagella antisera and monovalent flagella antisera for traditional slide agglutination testing.