Lysing Matrix E, MP Biomedicals
Supplier: MP Biomedicals
116914500 116914050 116914100 116984001 116984010
IC116914500PK 1971.02 USD
IC116914500 75784-628 75784-630 75856-180 75856-384
Lysing Matrix E, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix E is effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
  • Impact resistant tube
  • Contains 1.4mm ceramic spheres, 0.1mm silica spheres and 4mm glass beads
  • DNase and RNase Free
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under aMore Product Information