Lysing Matrix A, MP Biomedicals

Supplier: MP Biomedicals
116910500 116910050 116910100
IC116910500PK 1857.81 USD
IC116910500 75784-610 75784-612
Lysing Matrix A, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA.

  • Chemically inert
  • Will not bind nucleic acids
  • Used for all sample types except soil

Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA

Each impact-resistant 2 mL tube contains garnet matrix and one, 1/4 inch ceramic sphere. Extra 1/4 inch ceramic spheres are packaged separately. Lysing Matrix A tubes have orange caps and are found in the FastDNA™ and FastRNA™ SPIN Kits. Lysing Matrix A is used forMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Tube Compatibility 2 mL 2 ml
Specific Gravity Zirconium sphere, 5.5 - 6.1; Garnet Flakes, 4.0 - 4.1 Zirconium sphere, 5.5 - 6.1; Garnet Flakes, 4.0 - 4.1
Sample Volume up to 200 mg per prep up to 200 mg per prep
Storage and Handling Store at Room Temperature (15-30 °C) Store at Room Temperature (15...30 °C)
Sample Type Skeletal Muscle; Lung; Heart; Bone; Seeds; Bacteria (Gram +/-); Yeast; Fungi; Spores; Plants; Fixed Samples; Sludge; Wastewater; Feces; Food; Roots; Soil; Animal Tissue Skeletal Muscle; Lung; Heart; Bone; Seeds; Bacteria (Gram +/-); Yeast; Fungi; Spores; Plants; Fixed Samples; Sludge; Wastewater; Feces; Food; Roots; Soil; Animal Tissue
Product Type Lab Essentials Lab Essentials
Molecule Enriched DNA; RNA; Proteins; Other small molecules DNA; RNA; Proteins; Other small molecules
Matrix Type A A
Materials Required (Not Supplied) FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
Material of Construction Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere. Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere.
Key Applications Extractions of nucleic acids, proteins, metabolites and other small molecules from virtually any sample Extractions of nucleic acids, proteins, metabolites and other small molecules from virtually any sample
Instrument Compatibility FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2 FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
Adapter Compatibility QuickPrep (24x2 mL); HiPrep (48x2 mL); CoolPrep (24x2 mL); QuickFlex (96x2 mL); ConeFlex Legacy (allows all FP24 adapters on FP96) QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
Application Areas Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR