Corning® Ice Pan and Buckets, Corning
Supplier: Corning
432093 432094 432095 432096 432097 432098 432099 432100 432101 432102 432103 432104 432105 432106 432107 432108 432109 432110 432111 432112 432113 432114 432115 432116 432117 432118 432119 432120 432121 432122 432123 432124 432125 432126 432127 432128 432129 432130 432131 432132 432133 432134 432135
75779-804CS 209.38 USD
75779-804 75779-902 75779-904 75779-906 75779-908 75779-910 75779-912 75779-914 75779-916 75779-918 75779-920 75779-922 75779-924 75779-926 75779-928 75779-930 75779-932 75779-934 75779-936 75779-938 75779-940 75779-946 75779-948 75779-950 75779-952 75779-954 75779-956 75779-958 75779-960 75779-962 75779-964 75779-966 75779-968 75779-970 75779-972 75779-974 75779-976 75779-978 75779-980 75779-982 75779-984 75779-986 75779-988
Corning® Ice Pan and Buckets, Corning
Ice Containers
Ethylene vinyl acetate (EVA) foam.
  • Solid, one piece moulded, unbreakable
  • Resistant to highly acidic and basic solutions, salts, alcohol, acetone, chloroform, phenol, etc.
  • Insulative properties and resistant to temperatures from −196 to 93 °C
Ideal for use with ice, dry ice, liquid nitrogen, saline solution and acetone.
Corning ice pans and buckets are multi-purpose pans and buckets that are a great non-toxic alternative to conventional ice pans.
EVA has little or no odor, is inert and used in biomedical engineering applications such as drugMore Product Information