B.O.D. Bottles, Bar-Coded, 300 ml, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
227497-01 227497-02 227497-03 227497-00 227497-04 227497-05 227497-06 227497-09 227497-12 227497-13 227497-14 227497-15 227497-16 227497-18 227497-00G 227497-01G 227497-02G 227497-03G 227497-04G 227497-05G 227497-06G 227497-07G 227497-08G 227497-09G 227497-10G 227497-11G 227497-12G 227497-13G 227497-14G 227497-15G 227497-16G 227497-17G 227497-18G 227498 227497-07 227497-08 227497-10 227497-11 227497-17
16285-114CS 572.93 USD
16285-114 16285-125 16285-136 16285-140 16285-147 16285-158 16285-169 16285-191 16285-197 16285-199 16285-201 16285-203 16285-205 16285-209 74830-152 74830-154 74830-156 74830-158 74830-160 74830-162 74830-164 74830-166 74830-168 74830-170 74830-172 74830-174 80093-752 80093-754 80093-756 80093-758 80093-760 80093-762 80093-764 80093-766 16285-170 16285-180 16285-193 16285-195 16285-207
B.O.D. Bottles, Bar-Coded, 300 ml, WHEATON®, DWK Life Sciences
Bottles BOD Bottles
Incubation bottles are ideal for determining oxygen demand of sewage, water, and industrial wastes.
Borosilicate glass bottles are designed with large permanently screened-on labels for accuracy in recording bottle numbers. Interchangeable ground glass robotic stoppers and glass pennyhead stoppers are tapered to prevent air entrapment. Bottles fit most dissolved oxygen probes.
Diluted samples may be analyzed according to Standard Methods for the Examination of Water and Wastewater, 19th Ed., 5210B.
Order Now
SPECIFICATIONS
Environmentally Preferable