Nunc® Immobilizer™ Glutathione, Nickel-Chelate, and Streptavidin Plates and Modules, Thermo Scientific
Supplier: Thermo Fisher Scientific
436014 436022 436015 436020 436016
73521-130CS 712.22 USD
73521-130 73521-134 73521-414 73521-132 73521-416
Nunc® Immobilizer™ Glutathione, Nickel-Chelate, and Streptavidin Plates and Modules, Thermo Scientific
Microplates Immunology Microplates and Strips
Choose from solid polystyrene plates in a standard 96-well format or polystyrene modules consisting of twelve 8-well strips
All of the plates utilize spacer arm chemistry to covalently couple the affinity capture surface molecules. The glutathione plates are ideal for rapid coupling of GST-tagged fusion proteins. Nickel-chelate plates are ideal for rapid coupling of 6xHistidine-tagged proteins. Streptavidin plates are ideal for binding of biotinylated biomolecules such as peptides, antibodies, nucleic acids, or haptens and have an excellent binding capacity of 20pmol/mL.
Choose clearMore Product Information