Seg-Safe™ Segment Processor, Alpha Laboratories
Supplier: Alpha Scientific
201
73000-079CS 2892.55 USD
73000-079
Seg-Safe™ Segment Processor, Alpha Laboratories
Pipettes Pasteur/Transfer Pipettes
Ideal for blood typing and cross matching.
The processor samples blood bank segments and delivers calibrated drops without requiring scissors, slitters, and transfer pipettes. Diluent may be added directly to the processor, eliminating the need to dilute blood in a separate test tube. The single-use device consists of a protected stainless steel cannula for puncturing the segment, a soft plastic body for holding the cell suspension, and a spout for dispensing calibrated drops. Compatible with standard centrifuges for 12×75 mm test tubes. Non sterile.