Fast-Freeze® Flasks and Adapters, Labconco®
Supplier: Labconco
7540000 7542800 7540200 7540300 7540400 7540600 7540800 7540900 7541000 7541200 7545000 7545600 7545200 7545400 7547000 7547200 7545800 7546000 7547400 7547600 7542900 7543000 7544200 7544400 7541100 7543300 7543400 7544000 7542000 7542200 7542300 7542400 7542600 7543200 7542700 7540601 7540801 7540901 7541101 7542601 7542701 7542801 7542901
26675-228EA 137 USD
26675-228 26675-230 26675-240 26675-250 26675-261 26675-283 26675-308 26675-319 26675-320 26675-341 26675-352 26675-355 26675-363 26675-374 26675-385 26675-396 26675-443 26675-487 26675-501 26675-512 72460-048 72460-050 74920-002 74920-004 82018-226 89187-294 89187-296 72460-036 72460-038 72460-040 72460-042 72460-044 72460-046 72460-052 10148-032 75841-038 75841-040 75841-042 75841-044 75841-046 75841-048 75841-050 75841-052
Fast-Freeze® Flasks and Adapters, Labconco®
Flasks Flat Bottom Flasks
Fast-Freeze® Flasks for FreeZone® Freeze Dry Systems are designed to be easier to handle, faster to load, and more convenient to use in the laboratory for freeze drying.
  • Compatible with all major brands of freeze drying equipment, including Labconco and Virtis™
Fast-Freeze® Flasks simplify lyophilization procedures with a wide-mouth design that allows easy access to samples
Flat bottom, high-strength borosilicate glass flask withstands high heat and deep vacuum. Uniform glass walls maximize heat transfer while maintaining structural strength. Wide-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable