Nalgene® Amber HDPE Carboy, Thermo Scientific
Supplier: Thermo Fisher Scientific
2256-7020
71490-028CS 825.86 USD
71490-028
Nalgene® Amber HDPE Carboy, Thermo Scientific
Bottles Carboys
Protect photosensitive chemicals, reagents, buffers, and standards when mixing and storing.
The Nalgene™ amber HDPE carboy is supplied with amber polypropylene closure, and meets the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.