Ammonia solution 29%, Cleanroom® MB MB Electronic_industry

Supplier: KMG
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

63925
71009-320CS 360.09 USD
71009-320
Ammonia solution 29%, Cleanroom® MB MB Electronic_industry
Ammonia solution

Ammonium hydroxide is used in a SC-1 solution to remove organics and particles from the surface of a wafer. The ammonium hydroxide allows undercutting of the silicon dioxide freeing particles from the wafer by lift-off.

Storage: 0⁰ F - 80⁰F

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Assay (NH3) 28.00 to 30.00 %
Color 10 APHA
Appearance Conforms
Substance Reducing KMnO4 Conforms
Carbon Dioxide (CO2) 10,000 ppb
Chloride (Cl) 200 ppb
Nitrate (NO3) 400 ppb
Phosphate (PO4) 100 ppb
Sulfate (SO4) 200 ppb
Aluminum (Al) 10.0 ppb
Antimony (Sb) 5.0 ppb
Arsenic (As) 5.0 ppb
Barium (Ba) 10.0 ppb
Beryllium (Be) 10.0 ppb
Bismuth (Bi) 10.0 ppb
Boron (B) 10.0 ppb
Cadmium (Cd) 10.0 ppb
Calcium (Ca) 10.0 ppb
Chromium (Cr) 10.0 ppb
Cobalt (Co) 10.0 ppb
Copper (Cu) 10.0 ppb
Gallium (Ga) 10.0 ppb
Germanium (Ge) 10.0 ppb
Gold (Au) 10.0 ppb
Iron (Fe) 10.0 ppb
Lead (Pb) 10.0 ppb
Lithium (Li) 10.0 ppb
Magnesium (Mg) 10.0 ppb
Manganese (Mn) 10.0 ppb
Molybdenum (Mo) 10.0 ppb
Nickel (Ni) 10.0 ppb
Niobium (Nb) 10.0 ppb
Potassium (K) 10.0 ppb
Silicon (Si) 10.0 ppb
Silver (Ag) 10.0 ppb
Sodium (Na) 10.0 ppb
Strontium (Sr) 10.0 ppb
Tantalum (Ta) 10.0 ppb
Thallium (Tl) 10.0 ppb
Tin (Sn) 10.0 ppb
Titanium (Ti) 10.0 ppb
Vanadium (V) 10.0 ppb
Zinc (Zn) 10.0 ppb
Zirconium (Zr) 10.0 ppb
0.5μ Particle Count 25 par/ml
1.0μ Particle Count 10 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR