SP Bel-Art Flaskup™ Flask Holders, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F38951-2010 F38951-2012 F38951-2006 F38951-2004 F38951-2002
11215-550PK 56.98 USD
11215-550 70680-088 13196-080 70680-091 70680-093 70680-097 70680-600
SP Bel-Art Flaskup™ Flask Holders, Bel-Art Products, a part of SP
Supports Flask Supports
These unique holders are perfect for keeping round and tapered bottom flasks upright.
  • Polypropylene construction
  • Durable and easy to clean
  • Autoclavable
  • Slight taper on inside wall
  • Open bottom to accommodate multiple flask sizes
The inside wall has a slight taper to the open bottom, allowing a variety of flask shapes and sizes to fit into one holder.
The wide base keeps flasks stable and side slots can be used as sight windows. Molded from polypropylene, they are durable and easy to clean. Autoclavable at 121°C (250°F).
Ordering information: More Product Information