Cinnamon leaf oil Ceylon Type

Supplier: SPECTRUM CHEMICAL MFG CORP

C1574-22.5KG C1574-100GM C1574-2.5KG C1574-500GM
700010-530EA 3934.55 USD
700010-530 700010-526 700010-528 700010-532
Cinnamon leaf oil Ceylon Type
Cinnamon leaf oil

Cinnamon Leaf Oil, Ceylon Type, from the bark of the Cinnamomum cassia tree, is used mainly as a spice and a flavoring agent for some desserts, confectionery, and pastries.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00130805
CAS Number: 8015-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR