Tropaeolin O

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: CI 14270, Sodium 2,4-Dihydroxyazobenzene-4'-sulfonate, Tropaeolin O sodium salt, Sulpho orange, Chrysoin, Sulfo orange, 2,4-Dihydroxyazobenzene-4'-sulphonic acid sodium salt, Sodium 2,4-Dihydroxyazobenzene-4'-sulphonate, 2,4-Dihydroxyazobenzene-4'-sulfonic acid sodium salt, Acid Orange 6, Resorcinol Yellow

TR145-100GM TR145-25GM
700010-154EA 294.4 USD
700010-154 700010-156
Tropaeolin O
Tropaeolin O

Tropaeolin O, also known as Chrysoine resorcinol or Yellow T, is a synthetic azo dye used as a pH indicator.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₉N₂O₅SNa
Storage Temperature: Ambient
MDL Number: MFCD00007499
CAS Number: 547-57-9

Order Now

Specification Test Results

Absorption Max 387 NM
Visual Transition Intervals pH 11.0 (Yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR