D-γ-Tocopherol ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (R)-2,7,8-Trimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, γ-Tocopherol

T1327-100MG
700009-388EA 340.7 USD
700009-388
D-γ-Tocopherol ≥95%
D-γ-Tocopherol

D-gamma-Tocopherol, also known as γ-Tocopherol, is considered to be Vitamin E. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₂₈H₄₈O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00066529
CAS Number: 54-28-4

Order Now

Specification Test Results

Assay Approx. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR