Creatine (N-amidinosarcosine) monohydrate ≥99% suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate

CR105-500GM CR105-50KGBL CR105-25GM CR105-100GM
700007-906EA 264.6 USD
700007-906 700007-908 700007-904 700007-900
Creatine (N-amidinosarcosine) monohydrate ≥99% suitable for use as excipient
Creatine (N-Amidinosarcosine) monohydrate

Creatine, Monohydrate may be used as a supplement to study the treatment of neuromuscular diseases, where it may aid in neuroprotection and by improving the cellular bioenergetic state.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₉N₃O₂·1H₂O
MW: 149.15 g/mol (131.13 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00071582
CAS Number: 6020-87-7
EINECS: 200-306-6

Order Now

Specification Test Results

Assay (%) 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR