DL-Tyrosine ≥98.5% for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH

T3072-25GM
700007-494EA 125.5 USD
700007-494
DL-Tyrosine ≥98.5% for biochemistry
DL-Tyrosine

DL-Tyrosine - Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₁NO₃
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00063074
CAS Number: 556-03-6
EINECS: 209-113-1
Merck Index: 12,09970

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR